Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSEN34
Entrez Gene ID
693919
Gene Name
tRNA splicing endonuclease subunit 34
Gene Aliases
-
Chromosome Location
Chr.19: 53951304 - 53955195 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261765.1 NP_001248694.1 - 836 64
XM_015124801.2 XP_014980287.1 - 875 64
XM_028838347.1 XP_028694180.1 - 870 64
XM_028838348.1 XP_028694181.1 - 948 64
XM_028838349.1 XP_028694182.1 - 1222 64
XM_028838350.1 XP_028694183.1 - 1226 64
GenBank mRNA JU336341.1 - - 773 64
JU477165.1 - - 915 64
JU477166.1 - - 837 64
JV045026.1 - - 836 64
JV601605.1 - - 938 64

View Full Product Details