Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCEANC2
Entrez Gene ID
714992
Gene Name
transcription elongation factor A N-terminal and central domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 170318518 - 170363721 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266853.1 NP_001253782.1 - 426 68
GenBank mRNA JU332910.1 - - 263 68
JU474306.1 - - 426 68

View Full Product Details