Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIPK1A
Entrez Gene ID
706803
Gene Name
divergent protein kinase domain 1A
Gene Aliases
FAM69A
Chromosome Location
Chr.1: 131215909 - 131337340 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266885.1 NP_001253814.1 - 544 94
GenBank mRNA JU334451.1 - - 492 94
JU475542.1 - - 544 94
JV468120.1 - - 521 94
JV616274.1 - - 550 94
JV683580.1 - - 526 94

View Full Product Details