The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012671.2 | NP_036803.1 | 4 - 5 | 504 | 70 |
XM_006236822.2 | XP_006236884.1 | 4 - 5 | 1397 | 70 | |
XM_008763049.1 | XP_008761271.1 | 4 - 5 | 1401 | 70 | |
XM_008763050.1 | XP_008761272.1 | 4 - 5 | 1398 | 70 | |
GenBank mRNA | KF366251.1 | - | 4 - 5 | 360 | 70 |
M31076.1 | - | 4 - 5 | 504 | 70 | |
X02004.1 | - | 5 - 6 | 504 | 70 |