Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pou4f3
Entrez Gene ID
364855
Gene Name
POU class 4 homeobox 3
Gene Aliases
-
Chromosome Location
Chr.18: 36713869 - 36716461 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108889.1 NP_001102359.1 1 - 2 122 70

View Full Product Details