Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ddx50
Entrez Gene ID
361848
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 50
Gene Aliases
-
Chromosome Location
Chr.20: 32239748 - 32269145 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013198.2 NP_001013216.2 4 - 5 727 103

View Full Product Details