The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190236.1 | NP_001177165.1 | 14 - 15 | 1715 | 65 |
NM_001190237.1 | NP_001177166.1 | 14 - 15 | 1712 | 65 | |
NM_001190238.1 | NP_001177167.1 | 13 - 14 | 1703 | 65 | |
NM_013013.2 | NP_037145.2 | 13 - 14 | 1706 | 65 | |
XM_006256403.1 | XP_006256465.1 | 14 - 15 | 1724 | 65 | |
GenBank mRNA | BC061759.1 | - | 13 - 14 | 1637 | 65 |
DY313840.1 | - | 4 - 5 | 571 | 65 | |
FQ216707.1 | - | 2 - 3 | 198 | 65 | |
FQ217536.1 | - | 13 - 14 | 1660 | 65 |