The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031726.1 | NP_113914.1 | 4 - 5 | 408 | 66 |
GenBank mRNA | AF240784.1 | - | 4 - 5 | 408 | 66 |