Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsd11b2
Entrez Gene ID
25117
Gene Name
hydroxysteroid 11-beta dehydrogenase 2
Gene Aliases
-
Chromosome Location
Chr.19: 37476083 - 37481326 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017081.2 NP_058777.1 1 - 2 386 100
GenBank mRNA BC087023.1 - 1 - 2 373 100
U22424.1 - 1 - 2 387 100

View Full Product Details