The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033701.1 | NP_001028873.1 | - | 811 | 71 |
XM_006234144.2 | XP_006234206.2 | - | 1171 | 71 | |
XM_006234145.2 | XP_006234207.1 | - | 1691 | 71 | |
XM_006234147.2 | XP_006234209.1 | 5 - 6 | 1888 | 71 | |
XM_008761747.1 | XP_008759969.1 | - | 937 | 71 | |
XM_008761748.1 | XP_008759970.1 | - | 1170 | 71 | |
GenBank mRNA | AM084708.1 | - | - | 811 | 71 |