Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc30a8
Entrez Gene ID
299903
Gene Name
solute carrier family 30 (zinc transporter), member 8
Gene Aliases
ZnT-8
Chromosome Location
Chr.7: 91832988 - 91868406 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130538.1 NP_001124010.1 3 - 4 728 67
XM_006241645.2 XP_006241707.1 3 - 4 728 67
XM_008765500.1 XP_008763722.1 3 - 4 485 67

View Full Product Details