Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ugt1a6
- Entrez Gene ID
-
113992
- Gene Name
- UDP glucuronosyltransferase 1 family, polypeptide A6
- Gene Aliases
-
UDPGT 1-6, Udpgt, Ugt1, Ugt1a7
- Chromosome Location
-
Chr.9: 95241609 - 95302822 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
J05132.1 |
- |
- |
|
72 |
- Gene Symbol
- Fabp1
- Entrez Gene ID
-
24360
- Gene Name
- fatty acid binding protein 1, liver
- Gene Aliases
-
Fabplg, L-FABP, SCP, SCP., p14
- Chromosome Location
-
Chr.4: 99063181 - 99066957 on Build Rnor_6.0
View Full Product Details