Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klkb1
Entrez Gene ID
25048
Gene Name
kallikrein B, plasma 1
Gene Aliases
KALP15, Kalp1, Klk3
Chromosome Location
Chr.16: 50151128 - 50175407 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012725.2 NP_036857.2 2 - 3 119 149
XM_006253121.2 XP_006253183.1 2 - 3 1053 149
GenBank mRNA BC089815.1 - 2 - 3 119 149
M30282.1 - 2 - 3 82 149
M58590.1 - 2 - 3 82 149

View Full Product Details