The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012730.1 | NP_036862.1 | 4 - 5 | 805 | 64 |
GenBank mRNA | AB008423.1 | - | 4 - 5 | 681 | 64 |
BC078897.1 | - | 4 - 5 | 740 | 64 | |
M16655.1 | - | 4 - 5 | 731 | 64 | |
M22330.1 | - | 4 - 5 | 805 | 64 |