The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012921.1 | NP_037053.1 | 3 - 4 | 1117 | 92 |
XM_006241293.2 | XP_006241355.1 | 4 - 5 | 929 | 92 | |
GenBank mRNA | BC158591.1 | - | 3 - 4 | 893 | 92 |
L14018.1 | - | 3 - 4 | 1117 | 92 |