The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013098.2 | NP_037230.2 | 3 - 4 | 516 | 81 |
GenBank mRNA | BC090067.1 | - | 3 - 4 | 516 | 81 |
D78592.1 | - | 3 - 4 | 528 | 81 | |
L37333.1 | - | 3 - 4 | 480 | 81 | |
U07993.1 | - | 3 - 4 | 478 | 81 |