Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpa1
Entrez Gene ID
24269
Gene Name
carboxypeptidase A1 (pancreatic)
Gene Aliases
-
Chromosome Location
Chr.4: 57952982 - 57959109 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016998.3 NP_058694.2 2 - 3 460 100
GenBank mRNA FQ232505.1 - 2 - 3 168 100
J00713.1 - 2 - 3 460 100
V01232.1 - 2 - 3 163 100

View Full Product Details