The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017194.1 | NP_058890.1 | 8 - 9 | 962 | 87 |
XM_006246678.2 | XP_006246740.1 | 9 - 10 | 1080 | 87 | |
XM_008767807.1 | XP_008766029.1 | 9 - 10 | 1063 | 87 | |
XM_008767808.1 | XP_008766030.1 | 10 - 11 | 1729 | 87 | |
GenBank mRNA | X13252.1 | - | 8 - 9 | 962 | 87 |
X74836.1 | - | 8 - 9 | 962 | 87 |