The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017225.2 | NP_058921.1 | 1 - 2 | 137 | 84 |
XM_006247125.2 | XP_006247187.1 | 1 - 2 | 205 | 84 | |
GenBank mRNA | AF040261.1 | - | 1 - 2 | 81 | 84 |
BC078854.1 | - | 1 - 2 | 142 | 84 |