Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ednrb
Entrez Gene ID
50672
Gene Name
endothelin receptor type B
Gene Aliases
Ednra, Etb
Chromosome Location
Chr.15: 88004775 - 88036354 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017333.1 NP_059029.1 7 - 8 1448 151
XM_006252431.2 XP_006252493.1 7 - 8 1729 151
GenBank mRNA S65355.1 - 7 - 8 1448 151
X57764.1 - 6 - 7 1397 151

View Full Product Details