Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt3
Entrez Gene ID
25731
Gene Name
synaptotagmin III
Gene Aliases
SIII
Chromosome Location
Chr.1: 100379220 - 100407862 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019122.1 NP_061995.1 2 - 3 250 88
XM_006228978.2 XP_006229040.1 3 - 4 708 88
XM_006228979.2 XP_006229041.1 3 - 4 713 88
XM_008759324.1 XP_008757546.1 4 - 5 438 88
XM_008759325.1 XP_008757547.1 3 - 4 342 88
GenBank mRNA AF375464.1 - 1 - 2 148 88
BC088145.1 - 4 - 5 463 88
D28512.1 - 2 - 3 250 88
EU441216.1 - 1 - 2 153 88

View Full Product Details