The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019189.2 | NP_062062.2 | 3 - 4 | 542 | 67 |
XM_006231748.2 | XP_006231810.1 | 3 - 4 | 743 | 67 | |
XM_008760634.1 | XP_008758856.1 | 2 - 3 | 952 | 67 | |
GenBank mRNA | BC128740.1 | - | 3 - 4 | 542 | 67 |