The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019359.1 | NP_062232.1 | 6 - 7 | 881 | 91 |
GenBank mRNA | BC062020.1 | - | 6 - 7 | 884 | 91 |
FQ209647.1 | - | 1 - 2 | 104 | 91 | |
FQ228936.1 | - | 3 - 4 | 318 | 91 | |
U06755.1 | - | 6 - 7 | 881 | 91 |