The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145771.2 | NP_665714.2 | 4 - 5 | 347 | 96 |
XM_008765674.1 | XP_008763896.1 | 4 - 5 | 531 | 96 | |
GenBank mRNA | AF197128.1 | - | 4 - 5 | 341 | 96 |
AF230096.1 | - | 3 - 4 | 349 | 96 | |
AY738259.1 | - | 3 - 4 | 341 | 96 | |
BC078840.1 | - | 4 - 5 | 347 | 96 | |
FQ219187.1 | - | 3 - 4 | 336 | 96 |