Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem33
Entrez Gene ID
59303
Gene Name
transmembrane protein 33
Gene Aliases
-
Chromosome Location
Chr.14: 42537352 - 42559580 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034198.2 NP_001029370.1 2 - 3 203 83
NM_021671.3 NP_067703.1 2 - 3 203 83
GenBank mRNA AB006135.1 - 2 - 3 161 83
CK359233.1 - 2 - 3 84 83

View Full Product Details