The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022211.1 | NP_071547.1 | 2 - 3 | 459 | 137 |
GenBank mRNA | D64085.1 | - | 2 - 3 | 459 | 137 |