The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022392.1 | NP_071787.1 | 4 - 5 | 792 | 68 |
GenBank mRNA | BC078827.1 | - | 4 - 5 | 802 | 68 |
L13619.1 | - | 4 - 5 | 792 | 68 |