The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024157.1 | NP_077071.1 | 4 - 5 | 674 | 64 |
XM_008761526.1 | XP_008759748.1 | 4 - 5 | 823 | 64 | |
GenBank mRNA | BC089798.1 | - | 4 - 5 | 679 | 64 |
Y18965.1 | - | 4 - 5 | 674 | 64 |