The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031017.1 | NP_112279.1 | 8 - 9 | 922 | 109 |
NM_134443.1 | NP_604392.1 | 6 - 7 | 841 | 109 | |
XM_006245065.2 | XP_006245127.1 | 8 - 9 | 928 | 109 | |
XM_006245066.2 | XP_006245128.1 | 7 - 8 | 1093 | 109 | |
XM_006245067.2 | XP_006245129.1 | 7 - 8 | 833 | 109 | |
XM_006245068.2 | XP_006245130.1 | 6 - 7 | 780 | 109 | |
XM_006245069.2 | XP_006245131.1 | 6 - 7 | 1430 | 109 | |
XM_006245070.2 | XP_006245132.1 | 3 - 4 | 300 | 109 | |
XM_008767138.1 | XP_008765360.1 | 5 - 6 | 3284 | 109 | |
XM_008767139.1 | XP_008765361.1 | 2 - 3 | 1154 | 109 | |
GenBank mRNA | BC097311.1 | - | 9 - 10 | 1186 | 109 |
FQ211925.1 | - | 8 - 9 | 987 | 109 | |
X14788.1 | - | 8 - 9 | 922 | 109 | |
X60002.1 | - | 6 - 7 | 841 | 109 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008772050.1 | XP_008770272.1 | 1 - 1 | 876 | 109 |