The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031238.2 | NP_112517.1 | 3 - 4 | 528 | 147 |
XM_006229480.2 | XP_006229542.1 | 3 - 4 | 316 | 147 | |
XM_006229481.2 | XP_006229543.1 | 3 - 4 | 409 | 147 | |
XM_006229483.2 | XP_006229545.1 | 3 - 4 | 515 | 147 | |
GenBank mRNA | AF227439.1 | - | 3 - 4 | 531 | 147 |
BC078800.1 | - | 3 - 4 | 531 | 147 |