The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031622.2 | NP_113810.2 | 3 - 4 | 1460 | 70 |
XM_006243409.2 | XP_006243471.1 | 3 - 4 | 1954 | 70 | |
GenBank mRNA | BC109380.1 | - | 3 - 4 | 1411 | 70 |
M64301.1 | - | 2 - 3 | 1088 | 70 |