The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112712.1 | NP_001106183.1 | 13 - 14 | 1413 | 64 |
NM_001112713.1 | NP_001106184.1 | 13 - 14 | 1413 | 64 | |
NM_031657.3 | NP_113845.2 | 13 - 14 | 1413 | 64 | |
XM_006253653.2 | XP_006253715.1 | 13 - 14 | 1522 | 64 | |
XM_008771504.1 | XP_008769726.1 | 12 - 13 | 1680 | 64 | |
GenBank mRNA | AF040750.1 | - | 13 - 14 | 1413 | 64 |
X97439.1 | - | 13 - 14 | 1413 | 64 | |
X97440.1 | - | 13 - 14 | 1413 | 64 | |
Y09365.1 | - | 13 - 14 | 1441 | 64 |