The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031769.1 | NP_113957.1 | 5 - 6 | 382 | 63 |
GenBank mRNA | AF020346.1 | - | 5 - 6 | 382 | 63 |
FQ213832.1 | - | 5 - 6 | 486 | 63 |