Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vamp8
Entrez Gene ID
83730
Gene Name
vesicle-associated membrane protein 8
Gene Aliases
-
Chromosome Location
Chr.4: 100250301 - 100252756 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031827.1 NP_114015.1 1 - 2 162 73
GenBank mRNA AF132812.1 - 1 - 2 162 73

View Full Product Details