The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053526.2 | NP_445978.1 | 2 - 3 | 617 | 75 |
GenBank mRNA | AB042599.1 | - | 2 - 3 | 626 | 75 |
BC088124.1 | - | 2 - 3 | 617 | 75 |