Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arfrp1
Entrez Gene ID
117051
Gene Name
ADP-ribosylation factor related protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 176857667 - 176865105 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053980.1 NP_446432.1 1 - 2 128 64
GenBank mRNA X78603.1 - 1 - 2 128 64

View Full Product Details