The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133396.2 | NP_596887.1 | 2 - 3 | 572 | 92 |
XM_006238628.2 | XP_006238690.1 | 2 - 3 | 803 | 92 | |
GenBank mRNA | AB049402.1 | - | 2 - 3 | 573 | 92 |
BC128727.1 | - | 2 - 3 | 578 | 92 |