The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134458.2 | NP_604453.2 | 3 - 4 | 246 | 70 |
XM_006247249.2 | XP_006247311.1 | 3 - 4 | 1098 | 70 | |
GenBank mRNA | AF352812.1 | - | 3 - 4 | 246 | 70 |
CK840952.1 | - | 5 - 6 | 70 |