The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138862.3 | NP_620217.2 | 8 - 9 | 1081 | 121 |
XM_006240413.2 | XP_006240475.1 | 7 - 8 | 1064 | 121 | |
XM_006240414.2 | XP_006240476.1 | 8 - 9 | 1156 | 121 | |
XM_006240415.2 | XP_006240477.1 | 6 - 7 | 989 | 121 | |
XM_008764737.1 | XP_008762959.1 | 7 - 8 | 1035 | 121 | |
GenBank mRNA | BC078728.1 | - | 8 - 9 | 807 | 121 |