The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138905.2 | NP_620260.2 | 1 - 2 | 560 | 66 |
GenBank mRNA | BC072544.1 | - | 1 - 2 | 560 | 66 |
Y07783.1 | - | 1 - 2 | 552 | 66 |