Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd48
Entrez Gene ID
245962
Gene Name
Cd48 molecule
Gene Aliases
SLAMF2
Chromosome Location
Chr.13: 90116765 - 90141595 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139103.1 NP_620803.1 3 - 4 676 78
GenBank mRNA BC060542.1 - 3 - 4 713 78
FQ228817.1 - 3 - 4 730 78
FQ234539.1 - 3 - 4 78
X13016.1 - 3 - 4 676 78

View Full Product Details