The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139338.2 | NP_647554.1 | 12 - 13 | 1421 | 73 |
XM_006233558.2 | XP_006233620.1 | 13 - 14 | 1634 | 73 | |
XM_006233559.2 | XP_006233621.1 | 13 - 14 | 1750 | 73 | |
XM_006233560.2 | XP_006233622.1 | 13 - 14 | 1611 | 73 | |
XM_006233561.2 | XP_006233623.1 | 12 - 13 | 1524 | 73 | |
XM_006233562.2 | XP_006233624.1 | 12 - 13 | 1640 | 73 | |
XM_006233564.2 | XP_006233626.1 | 12 - 13 | 1504 | 73 | |
XM_006233565.2 | XP_006233627.1 | 13 - 14 | 1610 | 73 | |
XM_006233566.2 | XP_006233628.1 | 12 - 13 | 1616 | 73 | |
XM_006233567.2 | XP_006233629.1 | 12 - 13 | 1492 | 73 | |
XM_008761577.1 | XP_008759799.1 | 13 - 14 | 1699 | 73 | |
GenBank mRNA | AB077042.1 | - | 12 - 13 | 1426 | 73 |