The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144762.2 | NP_658907.1 | 1 - 2 | 457 | 129 |
GenBank mRNA | AB073318.1 | - | 1 - 2 | 457 | 129 |
AF371321.1 | - | 1 - 2 | 163 | 129 |