The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152936.1 | NP_690919.1 | 1 - 2 | 113 | 129 |
GenBank mRNA | FQ209906.1 | - | 1 - 2 | 94 | 129 |
FQ210000.1 | - | 1 - 2 | 127 | 129 | |
FQ210029.1 | - | 1 - 2 | 126 | 129 | |
FQ210105.1 | - | 1 - 2 | 131 | 129 | |
FQ210138.1 | - | 1 - 2 | 126 | 129 | |
FQ210442.1 | - | 1 - 2 | 126 | 129 | |
FQ210447.1 | - | 1 - 2 | 126 | 129 | |
FQ210766.1 | - | 1 - 2 | 128 | 129 | |
FQ210836.1 | - | 1 - 2 | 128 | 129 | |
FQ210933.1 | - | 1 - 2 | 128 | 129 | |
FQ211109.1 | - | 1 - 2 | 126 | 129 | |
FQ211214.1 | - | 1 - 2 | 130 | 129 | |
FQ211434.1 | - | 1 - 2 | 112 | 129 | |
FQ218107.1 | - | 1 - 2 | 128 | 129 | |
FQ218122.1 | - | 1 - 2 | 127 | 129 | |
FQ218165.1 | - | 1 - 2 | 128 | 129 | |
FQ218208.1 | - | 1 - 2 | 124 | 129 | |
FQ218422.1 | - | 1 - 2 | 126 | 129 | |
M27883.1 | - | 1 - 2 | 113 | 129 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003751771.3 | XP_003751819.1 | 1 - 2 | 463 | 129 |