Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Has1
Entrez Gene ID
282821
Gene Name
hyaluronan synthase 1
Gene Aliases
-
Chromosome Location
Chr.1: 59720612 - 59732409 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172323.1 NP_758826.1 4 - 5 1113 72
GenBank mRNA AB097568.1 - 4 - 5 1113 72

View Full Product Details