The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172334.2 | NP_758837.1 | 3 - 4 | 434 | 70 |
XM_006230701.2 | XP_006230763.1 | 3 - 4 | 490 | 70 | |
GenBank mRNA | AY093421.1 | - | 3 - 4 | 435 | 70 |
BC061802.1 | - | 3 - 4 | 455 | 70 | |
FQ228181.1 | - | 3 - 4 | 486 | 70 | |
FQ234469.1 | - | 3 - 4 | 486 | 70 | |
FQ235188.1 | - | 2 - 3 | 411 | 70 |