Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fabp3
Entrez Gene ID
79131
Gene Name
fatty acid binding protein 3, muscle and heart
Gene Aliases
-
Chromosome Location
Chr.5: 148528854 - 148535597 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024162.1 NP_077076.1 1 - 2 109 70
GenBank mRNA FQ217465.1 - 1 - 2 113 70
J02773.1 - 1 - 2 109 70
M18034.1 - 1 - 2 96 70

View Full Product Details