Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cltb
Entrez Gene ID
116561
Gene Name
clathrin, light chain B
Gene Aliases
-
Chromosome Location
Chr.17: 10537351 - 10554999 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053835.1 NP_446287.1 3 - 4 375 63
XM_006253595.2 XP_006253657.1 3 - 4 463 63
GenBank mRNA FQ213983.1 - 3 - 4 465 63
FQ229322.1 - 3 - 4 394 63
M15883.1 - 3 - 4 375 63
M19262.1 - 3 - 4 375 63

View Full Product Details