Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ap2m1
Entrez Gene ID
116563
Gene Name
adaptor-related protein complex 2, mu 1 subunit
Gene Aliases
Ap50, Clapm1, mu2
Chromosome Location
Chr.11: 84041184 - 84047542 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053837.1 NP_446289.1 8 - 9 1024 71
GenBank mRNA BC087724.1 - - 1031 71
FQ234012.1 - 3 - 4 286 71
M23674.1 - 8 - 9 1024 71

View Full Product Details