Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tagln
Entrez Gene ID
25123
Gene Name
transgelin
Gene Aliases
Sm22
Chromosome Location
Chr.8: 50222895 - 50228369 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031549.2 NP_113737.1 3 - 4 405 85
GenBank mRNA BC061770.1 - 3 - 4 405 85
FQ212757.1 - 3 - 4 430 85
FQ219883.1 - 3 - 4 428 85
FQ220131.1 - 3 - 4 429 85
FQ220322.1 - 3 - 4 427 85
FQ220861.1 - 3 - 4 430 85
FQ220863.1 - 3 - 4 430 85
FQ224487.1 - 3 - 4 430 85
FQ229263.1 - 3 - 4 428 85
FQ229406.1 - 3 - 4 427 85
FQ229775.1 - 3 - 4 429 85
FQ229807.1 - 3 - 4 430 85
FQ234912.1 - 3 - 4 425 85
M83107.1 - 3 - 4 515 85
X64422.1 - 2 - 3 353 85
X71070.1 - 3 - 4 402 85

View Full Product Details